2-Amino-3-fluorophenol 97%

Supplier: Ambeed
Warning

A197375-10G A197375-250MG A197375-1G A197375-5G
103816-912EA 41 USD
103816-912 103847-534 76999-206 76999-208 77085-538 77302-548
2-Amino-3-fluorophenol 97%
2-Amino-3-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₆FNO
MW: 127.1163 g/mol
Boiling Pt: 221.9 °C (760 mm Hg)
Melting Pt: 126…127 °C
Flash Pt: 88 °C
Storage Temperature: Freezer
MDL Number: MFCD07368754
CAS Number: 53981-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR