3-Fluoroanthranilic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-3-fluorobenzoic acid
A303335-10G
A303335-1G
A303335-25G
A303335-5G
103814-370EA
5
USD
103814-370
103835-538
76772-676
76838-222
76838-224
76838-226
3-Fluoroanthranilic acid 98%
3-Fluoroanthranilic acid
Precaución: for Research Use Only
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 294.4 °C (760 mm Hg) Melting Pt: 171…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569395 CAS Number: 825-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...