2-Amino-3-chlorophenol 98%

Supplier: Ambeed
Warning

A125153-100MG A125153-10G A125153-250MG A125153-25G A125153-1G
103800-632EA 27 USD
103800-632 76781-938 76781-940 76781-942 76781-944 77312-776
2-Amino-3-chlorophenol 98%
2-Amino-3-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.5700 g/mol
Flash Pt: 100.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11848418
CAS Number: 56962-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR