2-Amino-3-chlorobenzonitrile 98%

Supplier: Ambeed
Warning

A164807-100G A164807-100MG A164807-10G A164807-250MG A164807-1G A164807-25G A164807-5G
103805-906EA 13 USD
103805-906 103844-162 76768-766 76768-768 76768-770 76768-772 77320-348 77320-350 77320-352
2-Amino-3-chlorobenzonitrile 98%
2-Amino-3-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Boiling Pt: 266.9 °C (760 mm Hg)
Flash Pt: 115.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425881
CAS Number: 53312-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR