3-Chloroanthranilic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-chlorobenzoic acid

A154063-100G A154063-10G A154063-25G A154063-5G A154063-1000G
103842-778EA 5 USD
103842-778 77137-896 77137-898 77318-466 77318-468 77746-060
3-Chloroanthranilic acid 98%
3-Chloroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 316.3 °C (760 mm Hg)
Melting Pt: 189…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00134229
CAS Number: 6388-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR