2-Amino-3-chlorobenzaldehyde 98%

Supplier: Ambeed
Warning

A157626-100MG A157626-250MG
103805-028EA 45 USD
103805-028 103843-230 76973-388 76973-390
2-Amino-3-chlorobenzaldehyde 98%
2-Amino-3-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD13193431
CAS Number: 397322-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR