2-Amino-3-bromopyrazine 95%

Supplier: Ambeed
Warning

A320378-1G A320378-25G A320378-5G A320378-100MG A320378-10G A320378-250MG
103825-234EA 24 USD
103825-234 103836-168 76839-042 76839-044 76839-046 77251-116 77251-118 77251-120
2-Amino-3-bromopyrazine 95%
2-Amino-3-bromopyrazine

Precaución: for Research Use Only

Formula: C₄H₄BrN₃
Boiling Pt: 275.6 °C (760 mm Hg)
Flash Pt: 120.481 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909692
CAS Number: 21943-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR