2-Amino-3-bromophenol 97%

Supplier: Ambeed
Warning

A110975-100MG A110975-250MG A110975-1G A110975-5G A110975-25G A110975-10G
103797-768EA 20 USD
103797-768 103823-576 76781-062 76781-064 77310-382 77310-384 77570-624 77666-622
2-Amino-3-bromophenol 97%
2-Amino-3-bromophenol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
Boiling Pt: 266.2 °C (760 mm Hg)
Melting Pt: 138 °C
Flash Pt: 115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08445676
CAS Number: 116435-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR