2-Amino-3-bromo-6-methylpyridine 98%

Supplier: Ambeed
Warning

A133213-10G A133213-1G A133213-25G A133213-5G
103801-848EA 14 USD
103801-848 77109-376 77314-478 77314-480 77314-482
2-Amino-3-bromo-6-methylpyridine 98%
2-Amino-3-bromo-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
Melting Pt: 78…83 °C
Flash Pt: 98.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095215
CAS Number: 126325-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR