2-Amino-3-bromo-5-fluoropyridine 98%

Supplier: Ambeed
Warning

A323783-1G A323783-25G A323783-5G A323783-100G A323783-10G A323783-250MG
103825-358EA 8 USD
103825-358 103836-300 76839-212 76839-214 76839-216 76875-904 76875-906 76875-908
2-Amino-3-bromo-5-fluoropyridine 98%
2-Amino-3-bromo-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
Melting Pt: 62…66 °C
Flash Pt: 86.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03092924
CAS Number: 869557-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR