3-Bromo-5-fluoroanthranilic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-bromo-5-fluorobenzoic acid

A280105-100G A280105-10G A280105-250MG A280105-1G A280105-25G A280105-5G
103813-494EA 6 USD
103813-494 103853-438 77032-154 77032-156 77032-158 77093-368 77093-370 77093-372
3-Bromo-5-fluoroanthranilic acid 95%
3-Bromo-5-fluoroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
Boiling Pt: 344 °C (760 mm Hg)
Flash Pt: 161.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00662509
CAS Number: 259269-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR