2-Amino-3-benzyloxypyridine 98%

Supplier: Ambeed
Warning

A113746-100G A113746-10G A113746-25G A113746-500G A113746-1G A113746-5G
103798-288EA 5 USD
103798-288 103824-132 76766-022 76766-024 76766-026 76766-028 77297-266 77310-948
2-Amino-3-benzyloxypyridine 98%
2-Amino-3-benzyloxypyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
Boiling Pt: 361.8 °C (760 mm Hg)
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00006316
CAS Number: 24016-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR