2-Amino-3,5-difluoropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Difluoro-2-pyridylamine, 3,5-Difluoropyridin-2-amine

A241098-1G A241098-25G A241098-5G A241098-100G A241098-10G A241098-250MG
103811-392EA 18 USD
103811-392 103851-198 77090-358 77090-360 77090-362 77140-264 77140-266 77140-268
2-Amino-3,5-difluoropyridine 98%
2-Amino-3,5-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄F₂N₂
Boiling Pt: 155.8 °C (760 mm Hg)
Melting Pt: 55…57 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD03788760
CAS Number: 732306-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR