2-Amino-3,5-dibromobenzonitrile 97%

Supplier: Ambeed
Warning

A277264-1G A277264-25G A277264-5G A277264-100G A277264-10G
103813-344EA 5 USD
103813-344 103853-278 77093-170 77093-172 77093-174 77113-986 77113-988
2-Amino-3,5-dibromobenzonitrile 97%
2-Amino-3,5-dibromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂
Boiling Pt: 301.2 °C (760 mm Hg)
Melting Pt: 152…156 °C
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00179571
CAS Number: 68385-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR