3,5-Dibromo-6-chloropyrazin-2-amine 96%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3,5-dibromo-6-chloropyrazine

A101017-250MG A101017-1G A101017-5G
103795-756EA 40 USD
103795-756 103821-448 76780-442 77296-950 77296-952
3,5-Dibromo-6-chloropyrazin-2-amine 96%
3,5-Dibromo-6-chloropyrazin-2-amine

Precaución: for Research Use Only

Formula: C4H2Br2ClN3
MW: 287.3400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11043607
CAS Number: 566205-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR