2-Amino-3,4-dimethylbenzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-3,4-dimethylbenzoic Acid, ['3,4-Dimethylanthranilic acid']
A120582-100G
A120582-10G
A120582-25G
A120582-1G
A120582-5G
103799-960EA
5
USD
103799-960
76971-486
76971-488
76971-490
77297-774
77312-068
2-Amino-3,4-dimethylbenzoic acid 98%
2-Amino-3,4-dimethylbenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Boiling Pt: 341.7 °C (760 mm Hg) Melting Pt: 180 °C Flash Pt: 160.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130041 CAS Number: 50419-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...