2-Amino-3,4-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A110119-250MG A110119-5G
103797-620EA 41 USD
103797-620 103823-428 77245-652 77310-228
2-Amino-3,4-difluorobenzoic acid 97%
2-Amino-3,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Boiling Pt: 303.0±42.0 °C (760 mm Hg)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD09743511
CAS Number: 158580-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR