2-Amino-3,4-dichlorobenzoic acid 95%

Supplier: Ambeed
Warning

A101877-100MG A101877-250MG A101877-5G
103796-190EA 352 USD
103796-190 103821-884 77245-154 77245-156 77282-856
2-Amino-3,4-dichlorobenzoic acid 95%
2-Amino-3,4-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.02 g/mol
Boiling Pt: 356.2 °C (760 mm Hg)
Flash Pt: 169.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00078315
CAS Number: 20776-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR