2-Amino-3-(trifluoromethoxy)benzoic acid 98%

Supplier: Ambeed
Warning

A291820-5G A291820-1G A291820-25G
103835-060EA 46 USD
103835-060 76837-658 77032-308 77032-310
2-Amino-3-(trifluoromethoxy)benzoic acid 98%
2-Amino-3-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃
Boiling Pt: 282.8 °C (760 mm Hg)
Melting Pt: 99…101 °C
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD08669644
CAS Number: 561304-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR