3-(2-Pyridyl)-DL-alanine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(pyridin-2-yl)propionic acid, H-DL-β-(2-Pyridyl)-Ala-OH, H-β-(2-Pyridyl)-DL-Ala-OH

A160026-100MG A160026-250MG A160026-5G
103805-318EA 47 USD
103805-318 103843-548 77096-122 77096-124 77300-434
3-(2-Pyridyl)-DL-alanine 97%
3-(2-Pyridyl)-DL-alanine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 322.4 °C (760 mm Hg)
Flash Pt: 148.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00237828
CAS Number: 17407-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR