2-Chloro-DL-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-DL-phenylalanine, 2-Amino-3-(2-chlorophenyl)propanoic acid, 2-Chlorophenylalanine

A552450-1G A552450-25G A552450-5G A552450-100G A552450-10G
103828-698EA 10 USD
103828-698 103839-830 76843-906 76843-908 76843-910 77157-174 77157-176
2-Chloro-DL-phenylalanine 95%
2-Chloro-DL-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.6300 g/mol
Boiling Pt: 339.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00037774
CAS Number: 14091-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR