2-Amino-2-phenylbutanoic acid 97%

Supplier: Ambeed
Warning

A215779-1G A215779-5G A215779-10G A215779-100G
103818-514EA 12 USD
103818-514 103849-222 77087-772 77087-774 77249-216 77249-218 77615-812
2-Amino-2-phenylbutanoic acid 97%
2-Amino-2-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00038162
CAS Number: 5438-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR