AMP (2-Amino-2-methylpropanol) 97%

Supplier: Ambeed
Danger

Synonyms: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP

A101633-100G A101633-25G A101633-500G
103807-810EA 45 USD
103807-810 76994-524 76994-526 76994-528
AMP (2-Amino-2-methylpropanol) 97%
AMP (2-Amino-2-methylpropanol)

Precaución: for Research Use Only

Formula: (CH₃)₂C(NH₂)CH₂OH
MW: 89.14 g/mol
Boiling Pt: 165 °C (760 mm Hg)
Melting Pt: 24…28 °C
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00008051
CAS Number: 124-68-5
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR