2-Amino-2-cyclohexylacetic acid 97%
Supplier: Ambeed
|
Warning
|
A148809-100G
A148809-10G
A148809-250MG
A148809-1G
A148809-25G
A148809-5G
103803-966EA
6
USD
103803-966
103842-086
77110-186
77110-188
77110-190
77317-554
77317-556
77317-558
2-Amino-2-cyclohexylacetic acid 97%
2-Amino-2-cyclohexylacetic acid
Precaución: for Research Use Only
Formula:
C₈H₁₅NO₂ Boiling Pt: 292.8 °C (760 mm Hg) Melting Pt: 256 °C Flash Pt: 131 °C Storage Temperature: Ambient |
MDL Number:
MFCD05662746 CAS Number: 5664-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...