2-Amino-2-(4-methylphenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-(p-tolyl)acetic acid, H-DL-4-Me-Gly-OH

A193209-1G A193209-250MG
103816-512EA 40 USD
103816-512 103847-126 77085-024 77685-469
2-Amino-2-(4-methylphenyl)acetic acid 97%
2-Amino-2-(4-methylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 306.8 °C (760 mm Hg)
Melting Pt: 228 °C
Storage Temperature: Ambient
MDL Number: MFCD00665357
CAS Number: 13227-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR