Tris acetate 98%

Supplier: Ambeed
Warning

Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate

A511959-100G A511959-500G A511959-25G
103810-414EA 117 USD
103810-414 76790-018 76790-020 76843-490
Tris acetate 98%
Tris acetate

Precaución: for Research Use Only

Formula: NH₂C(CH₂OH)₃·CH₃COOH
MW: 181.1870 g/mol
Boiling Pt: 219…220 °C at 9.7513 mmHg
Melting Pt: 120…121 °C
Flash Pt: 169.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00038951
CAS Number: 6850-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR