2-Amino-2-(3-chlorophenyl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-DL-Phg(3-Cl)-OH

A164296-10G A164296-1G A164296-5G
103805-838EA 5 USD
103805-838 103844-098 77236-004 77320-256 77320-258
2-Amino-2-(3-chlorophenyl)acetic acid 95%
2-Amino-2-(3-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
Boiling Pt: 316.9 °C (760 mm Hg)
Melting Pt: 250…254 °C
Flash Pt: 145.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00049326
CAS Number: 7292-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR