2-Amino-4-hydroxypteridine 95%

Supplier: Ambeed
Warning

A239565-250MG A239565-1G A239565-5G
103811-300EA 130 USD
103811-300 103851-098 77000-374 77304-164 77304-166
2-Amino-4-hydroxypteridine 95%
2-Amino-4-hydroxypteridine

Precaución: for Research Use Only

Formula: C₆H₇N₅O
Boiling Pt: 424.7 °C (760 mm Hg)
Melting Pt: 300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01456481
CAS Number: 2236-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR