2-Amino-4'-fluoroacetophenone hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-1-(4-fluorophenyl)ethanone hydrochloride, 2-Amino-4'-fluoroacetophenone Hydrochloride, 4-Fluorophenacylamine hydrochloride

A271815-1G A271815-25G A271815-5G A271815-10G A271815-250MG
103813-066EA 32 USD
103813-066 103852-978 77092-762 77092-764 77092-766 77250-376 77250-378
2-Amino-4'-fluoroacetophenone hydrochloride 97%
2-Amino-4'-fluoroacetophenone hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉ClFNO
MW: 189.6146 g/mol
Boiling Pt: 296.6 °C (760 mm Hg)
Melting Pt: 235…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00193059
CAS Number: 456-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR