2-Allylisoindoline-1,3-dione 95%

Supplier: Ambeed
Warning

A162523-100MG A162523-10G A162523-250MG A162523-5G A162523-1G
103805-622EA 42 USD
103805-622 103843-872 76872-086 76872-088 76872-090 77300-596 77319-918
2-Allylisoindoline-1,3-dione 95%
2-Allylisoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
Boiling Pt: 296.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00158662
CAS Number: 5428-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR