2-Allyl-1-(6-(2-hydroxypropan-2-yl)pyridin-2-yl)-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidin-3(2H)-one 95%
Supplier: Ambeed
|
Warning
|
A272272-100MG
A272272-1G
103799-546EA
251
USD
103799-546
103813-086
76827-358
77305-102
2-Allyl-1-(6-(2-hydroxypropan-2-yl)pyridin-2-yl)-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidin-3(2H)-one 95%
2-Allyl-1-(6-(2-hydroxypropan-2-yl)pyridin-2-yl)-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidin-3(2H)-one
Precaución: for Research Use Only
Formula:
C₁₇H₁₉N₅O₂S MW: 357.43 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD19443207 CAS Number: 955369-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...