2-Acetylthiazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Thiazol-2-yl)-1-ethanone

A223052-25G A223052-100G A223052-10G A223052-1000G A223052-500G
103849-900EA 5 USD
103849-900 77088-516 77288-252 77288-254 77748-364 77748-365
2-Acetylthiazole 98%
2-Acetylthiazole

Precaución: for Research Use Only

Formula: C₅H₅NOS
MW: 127.17 g/mol
Boiling Pt: 212.5 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00005324
CAS Number: 24295-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR