2-Acetylphenanthrene 95%

Supplier: Ambeed
Warning

A298111-1G A298111-100MG A298111-250MG
103814-174EA 95 USD
103814-174 103835-318 76838-016 77032-432 77032-434
2-Acetylphenanthrene 95%
2-Acetylphenanthrene

Precaución: for Research Use Only

Formula: C₁₆H₁₂O
MW: 220.2700 g/mol
Boiling Pt: 405.9 °C (760 mm Hg)
Melting Pt: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00001171
CAS Number: 5960-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR