2-Acetylbenzo[b]thiophene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Acetylbenzothiophene
A126475-100G
A126475-10G
A126475-1G
A126475-5G
103800-816EA
11
USD
103800-816
76797-676
76797-678
77313-034
77313-036
2-Acetylbenzo[b]thiophene 98%
2-Acetylbenzo[b]thiophene
Precaución: for Research Use Only
Formula:
C₁₀H₈OS Boiling Pt: 304.5 °C (760 mm Hg) Melting Pt: 86…88 °C Flash Pt: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090217 CAS Number: 22720-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...