2-Acetyl-6-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(6-Methoxy-2-pyridinyl)ethanone

A153559-100MG A153559-10G A153559-250MG A153559-1G A153559-5G
103804-526EA 38 USD
103804-526 103842-700 77247-380 77247-382 77247-384 77300-010 77300-012
2-Acetyl-6-methoxypyridine 98%
2-Acetyl-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Boiling Pt: 214 °C (760 mm Hg)
Melting Pt: 40…44 °C
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD08276836
CAS Number: 21190-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR