2-Acetyl-6-bromopyridine 98%

Supplier: Ambeed
Warning

A115188-100G A115188-10G A115188-250MG A115188-25G A115188-500G A115188-1G A115188-5G
103798-528EA 9 USD
103798-528 103824-374 76982-922 76982-924 76982-926 76982-928 76982-930 77311-188 77311-190
2-Acetyl-6-bromopyridine 98%
2-Acetyl-6-bromopyridine

Precaución: for Research Use Only

Formula: C₇H₆BrNO
Boiling Pt: 271.2 °C (760 mm Hg)
Melting Pt: 51…55 °C
Flash Pt: 117.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00272200
CAS Number: 49669-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR