2-Acetyl-3-fluoropyridine 98%

Supplier: Ambeed
Warning

A102921-250MG A102921-25G A102921-1G A102921-5G
103796-448EA 14 USD
103796-448 103822-152 76869-208 76869-210 77309-022 77309-024
2-Acetyl-3-fluoropyridine 98%
2-Acetyl-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₇H₆FNO
MW: 139.1300 g/mol
Boiling Pt: 179.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06659495
CAS Number: 87674-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR