2-Acetamidoacrylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamidoacrylic acid

A152576-10G A152576-250MG A152576-25G A152576-5G
103804-406EA 27 USD
103804-406 103842-578 77247-324 77247-326 77299-962 77299-964
2-Acetamidoacrylic acid 97%
2-Acetamidoacrylic acid

Precaución: for Research Use Only

Formula: C₅H₇NO₃
MW: 129.12 g/mol
Boiling Pt: 418.5±37.0 °C (760 mm Hg)
Melting Pt: 185…186 °C
Flash Pt: 206.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004257
CAS Number: 5429-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR