2-Acetylamino-4-methylthiazole-5-sulfonyl chloride 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Acetylamino-4-methyl-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-1,3-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methylthiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-5-thiazolesulphonyl chloride
A125043-10G
A125043-1G
A125043-5G
103800-608EA
19
USD
103800-608
77246-280
77312-758
77312-760
2-Acetylamino-4-methylthiazole-5-sulfonyl chloride 97%
2-Acetylamino-4-methylthiazole-5-sulfonyl chloride
Precaución: for Research Use Only
Formula:
C₆H₇ClN₂O₃S₂ Melting Pt: 161…164 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005320 CAS Number: 69812-29-9 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...