2-Acetamido-2-cyclohexylacetic acid 97%

Supplier: Ambeed
Warning

A217790-10G A217790-1G A217790-100G
103818-656EA 26 USD
103818-656 103849-384 76986-938 77303-462 77683-628
2-Acetamido-2-cyclohexylacetic acid 97%
2-Acetamido-2-cyclohexylacetic acid

Precaución: For research use only.

Formula: C₁₀H₁₇NO₃
MW: 199.24 g/mol
Boiling Pt: 428.641 °C (760 mm Hg)
Flash Pt: 213.035 °C
Storage Temperature: Ambient
MDL Number: MFCD11046379
CAS Number: 107020-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR