20-(tert-Butoxy)-20-oxoicosanoic acid 97%

Supplier: Ambeed
Warning

A363139-10G A363139-250MG
103826-334EA 18 USD
103826-334 103837-354 77167-864 77167-866
20-(tert-Butoxy)-20-oxoicosanoic acid 97%
20-(tert-Butoxy)-20-oxoicosanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₄₆O₄
Storage Temperature: Ambient
MDL Number: MFCD23136041
CAS Number: 683239-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR