2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[N,N-Bis(Trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)triflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonyl)imide, Comins Triflating Reagent, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonimide)
A131286-100G
A131286-10G
A131286-1G
A131286-25G
A131286-5G
103801-556EA
5
USD
103801-556
77109-270
77109-272
77298-470
77314-046
77314-048
2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine 98%
2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine
Precaución: for Research Use Only
Formula:
C₇H₃ClF₆N₂O₄S₂ MW: 392.67 g/mol Boiling Pt: 358.3 °C (760 mm Hg) Melting Pt: 45…47 °C Flash Pt: 170.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00191833 CAS Number: 145100-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...