2-[2-(2-Propynyloxy)ethoxy]ethylamine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Propargyl-PEG2-amine, 2-[2-(Propargyloxy)ethoxy]ethylamine
A292945-1G
A292945-25G
A292945-5G
A292945-100G
A292945-10G
A292945-250MG
A292945-500G
103813-960EA
23
USD
103813-960
103835-100
76837-706
76837-708
76837-710
77099-356
77099-358
77099-360
77612-688
2-[2-(2-Propynyloxy)ethoxy]ethylamine 97%
2-[2-(2-Propynyloxy)ethoxy]ethylamine
Precaución: for Research Use Only
Formula:
C₇H₁₃NO₂ Boiling Pt: 213.8±20.0 °C (760 mm Hg) Flash Pt: 88.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD19160595 CAS Number: 944561-44-8 UN: 2735 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...