2,9-Dimethylquinolino[2,3-b]acridine-7,14(5H,12H)-dione Tinting strength > 99%

Supplier: Ambeed
Warning

A421029-1000G A421029-100G A421029-500G A421029-1G A421029-25G A421029-5G
103827-400EA 9 USD
103827-400 103838-446 76725-246 76725-248 76725-250 77307-208 77307-210 77307-212
2,9-Dimethylquinolino[2,3-b]acridine-7,14(5H,12H)-dione Tinting strength > 99%
2,9-Dimethylquinolino[2,3-b]acridine-7,14(5H,12H)-dione

Precaución: for Research Use Only

Formula: C₂₂H₁₆N₂O₂
MW: 340.39 g/mol
Boiling Pt: 601.6 °C (760 mm Hg)
Melting Pt: 440 °C (760 mm Hg)
Flash Pt: 221.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00071968
CAS Number: 980-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR