Bathocuproin 98%

Supplier: Ambeed
Warning

Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

A215244-100G A215244-10G A215244-250MG A215244-1G A215244-25G A215244-5G
103818-468EA 17 USD
103818-468 103849-172 76999-766 76999-768 76999-770 77087-696 77087-698 77087-700
Bathocuproin 98%
Bathocuproin

Precaución: for Research Use Only

Formula: C₂₆H₂₀N₂
MW: 360.46 g/mol
Melting Pt: 279…283 °C
Flash Pt: 231 °C
Storage Temperature: Ambient
MDL Number: MFCD00004972
CAS Number: 4733-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR