Neocuproine 98%

Supplier: Ambeed
Warning

Synonyms: 2,9-Dimethyl-1,10-phenanthroline

A199979-100G A199979-10G A199979-500G A199979-1G A199979-25G A199979-5G
103817-152EA 10 USD
103817-152 103847-778 76721-412 76721-414 76721-416 77302-636 77302-638 77302-640
Neocuproine 98%
Neocuproine

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Boiling Pt: 367.4 °C (760 mm Hg)
Melting Pt: 159…164 °C
Flash Pt: 159.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004973
CAS Number: 484-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR