2,9-Dibromo-1,10-phenanthroline 96%

Supplier: Ambeed
Warning

A122969-100MG A122969-250MG A122969-25G A122969-1G A122969-5G
103800-314EA 26 USD
103800-314 76810-616 76810-618 76810-620 77312-434 77312-436
2,9-Dibromo-1,10-phenanthroline 96%
2,9-Dibromo-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₂H₆Br₂N₂
Boiling Pt: 460.244 °C (760 mm Hg)
Flash Pt: 232.147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16038623
CAS Number: 39069-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR