2,8-Dichloroquinoline 98%

Supplier: Ambeed
Warning

A217772-1G A217772-25G A217772-5G A217772-10G A217772-250MG
103818-652EA 6 USD
103818-652 103849-380 77088-022 77088-024 77088-026 77249-294 77249-296
2,8-Dichloroquinoline 98%
2,8-Dichloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
Boiling Pt: 292.8 °C (760 mm Hg)
Melting Pt: 105…108 °C
Flash Pt: 158.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00832077
CAS Number: 4470-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR