2,8-Dibromodibenzofuran 97%

Supplier: Ambeed
Warning

Synonyms: 2,8-Dibromodibenzo[b,d]furan

A181798-100G A181798-10G A181798-250MG A181798-1G A181798-25G A181798-5G
103807-654EA 6 USD
103807-654 103846-026 76813-582 76813-584 76813-586 77083-550 77301-746 77301-748
2,8-Dibromodibenzofuran 97%
2,8-Dibromodibenzofuran

Precaución: for Research Use Only

Formula: C₁₂H₆Br₂O
MW: 325.99 g/mol
Boiling Pt: 396 °C (760 mm Hg)
Melting Pt: 226 °C
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00093683
CAS Number: 10016-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR