2,8-Diazaspiro[4.5]decane-1,3-dione hydrochloride 97%

Supplier: Ambeed
Warning

A352033-100MG A352033-250MG A352033-1G A352033-10G
103826-060EA 127 USD
103826-060 103837-056 77154-990 77154-992 77306-620 77686-000
2,8-Diazaspiro[4.5]decane-1,3-dione hydrochloride 97%
2,8-Diazaspiro[4.5]decane-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13185074
CAS Number: 2696-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR